missing translation for 'onlineSavingsMsg'
Learn More

3-(2,2-Dichloroethenyl)-2,2-dimethyl-cyclopropanecarboxylic Acid Anhydride (Mixture of Diastereomers), TRC

Product Code. 30136503
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136503 5 g 5g
30006310 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136503 Supplier Toronto Research Chemicals Supplier No. TRCD4342955G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56860-12-9
Molecular Formula C16 H18 Cl4 O3
Molecular Weight (g/mol) 400.12
Synonym Cyclopropanecarboxylic acid, 3-(2,2-dichloroethenyl)-2,2-dimethyl-, anhydride (9CI), 3-(2,2-Dichlorovinyl)-2,2-dimethylcyclopropanecarboxylic anhydride, 3-(2,2-Dichloroethenyl)-2,2-dimethyl-cyclopropanecarboxylic acid anhydride, 2-(2,2-Dichloroethenyl)-3,3-dimethylcyclopropane-1-carboxylic anhydride, Permethrin Imp. H (EP)
IUPAC Name [3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropanecarbonyl] 3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate
SMILES CC1(C)C(C=C(Cl)Cl)C1C(=O)OC(=O)C2C(C=C(Cl)Cl)C2(C)C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C16H18Cl4O3/c1-15(2)7(5-9(17)18)11(15)13(21)23-14(22)12-8(6-10(19)20)16(12,3)4/h5-8,11-12H,1-4H3
Formula Weight 398.001
Chemical Name or Material 2-(2,2-Dichloroethenyl)-3,3-dimethylcyclopropane-1-carboxylic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.