missing translation for 'onlineSavingsMsg'
Learn More

3-[2-(2-Aminoethylamino)ethylamino]propyl-trimethoxysilane, tech.

Product Code. 10686374 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10686374 500 mL Glass bottle 500mL
10143491 25 mL Glass bottle 25mL
10357901 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10686374 Supplier Thermo Scientific Acros Supplier No. 346175000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 35141-30-1 | C10H27N3O3Si | 265.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35141-30-1
Molecular Formula C10H27N3O3Si
Molecular Weight (g/mol) 265.43
MDL Number MFCD00069173
InChI Key NHBRUUFBSBSTHM-UHFFFAOYSA-N
Synonym n1-2-aminoethyl-n2-3-trimethoxysilyl propyl ethane-1,2-diamine, 3-trimethoxysilylpropyl diethylenetriamine, 3-2-2-aminoethylamino ethylamino propyl-trimethoxysilane, 3-trimethoxysilyl propyl diethylenetriamine, n1-3-trimethoxysilylpropyl diethylenetriamine, trimethoxysilylpropyldiethylenetriamine, 1,2-ethanediamine, n-2-aminoethyl-n'-3-trimethoxysilyl propyl, diethylenetriamino propyltrimethoxysilane, 3-2-2-aminoethylamino ethylamino propyltrimethoxysilane, n-2-aminoethyl-n'-3-trimethoxysilyl propyl ethylenediamine
PubChem CID 118771
IUPAC Name N'-[2-(3-trimethoxysilylpropylamino)ethyl]ethane-1,2-diamine
SMILES CO[Si](CCCNCCNCCN)(OC)OC

Specifications

Color Yellow
Density 1.0310g/mL
Boiling Point 114.0°C to 118.0°C (2.0 mmHg)
Flash Point 137°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula (H3CO)3Si(CH2)3NH(CH2)2NH(CH2)2NH2
Refractive Index 1.4570 to 1.4610
Quantity 500 mL
Specific Gravity 1.031
Solubility Information Solubility in water: immiscible
Formula Weight 265.43
Percent Purity 14 to 18% (amine (as NH2))
Grade Technical
Physical Form Liquid
Chemical Name or Material 3-[2-(2-Aminoethylamino)ethylamino]propyl-trimethoxysilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-[2-(2-Aminoethylamino)ethylamino]propyl-trimethoxysilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.