missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(1H-Tetrazol-5-yl)benzyl alcohol, 96%

Product Code. 11411620
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11411620 1 g 1g
15493577 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11411620 Supplier Alfa Aesar™ Supplier No. H26985.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 179057-20-6
Molecular Formula C8H8N4O
Molecular Weight (g/mol) 176.179
MDL Number MFCD09864536
InChI Key YGESRLFHDANLIL-UHFFFAOYSA-N
Synonym 3-1h-tetrazol-5-yl phenyl methanol, 3-1h-tetrazol-5-yl benzyl alcohol, 3-2h-tetrazol-5-yl phenyl methanol, 3-1h-1,2,3,4-tetrazol-5-yl phenyl methanol, 5-3-hydroxymethylphenyl-1h-tetrazole, benzenemethanol,3-2h-tetrazol-5-yl, 5-3-hydroxymethyl phenyl-2h-tetrazole, 3-2h-1,2,3,4-tetrazol-5-yl phenyl methanol
PubChem CID 23437784
IUPAC Name [3-(2H-tetrazol-5-yl)phenyl]methanol
SMILES C1=CC(=CC(=C1)CO)C2=NNN=N2

Specifications

Melting Point 208°C to 210°C
Quantity 1 g
Formula Weight 176.18
Percent Purity 96%
Chemical Name or Material 3-(1H-Tetrazol-5-yl)benzyl alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.