missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Tetrazol-1-yl)aniline, 97%, Thermo Scientific Chemicals

Product Code. 15420739
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14213-12-8
Molecular Formula C7H7N5
Molecular Weight (g/mol) 161.168
MDL Number MFCD06687920
InChI Key YXWXLBHIUVBLIH-UHFFFAOYSA-N
Synonym 3-1h-tetrazol-1-yl aniline, 3-tetrazol-1-yl-phenylamine, 3-1h-tetrazol-1-yl benzenamine, 3-1,2,3,4-tetrazol-1-yl aniline, 3-1h-1,2,3,4-tetrazol-1-yl aniline, 3-tetrazol-1-yl phenylamine, 3-1,2,3,4-tetraazolyl phenylamine, acmc-209cnn, 3-tetrazol-1-yl aniline, 3-tetrazol-1-ylphenylamine
PubChem CID 863007
IUPAC Name 3-(tetrazol-1-yl)aniline
SMILES C1=CC(=CC(=C1)N)N2C=NN=N2

Specifications

Quantity 5 g
Formula Weight 161.16
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-(1H-Tetrazol-1-yl)aniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'