missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Pyrrol-1-yl)propanenitrile, 97%, Thermo Scientific™

Product Code. p-4812854
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10673625 1 g 1g
10007014 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10673625 Supplier Thermo Scientific Maybridge Supplier No. CD01949DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 43036-06-2
Molecular Formula C7H8N2
Molecular Weight (g/mol) 120.16
MDL Number MFCD00003092
InChI Key IYOLJLGYJMJLSU-UHFFFAOYSA-N
Synonym 3-1h-pyrrol-1-yl propanenitrile, n-2-cyanoethyl pyrrole, 1-2-cyanoethyl pyrrole, pyrrole-1-propionitrile, 1h-pyrrole-1-propanenitrile, 3-pyrrol-1-yl propiononitrile, n-2-cyanoethyl-pyrrole, 3-pyrrol-1-yl propanenitrile, n-cyanoethylpyrrole, n-2-cyanoethyl pyrole
PubChem CID 96527
IUPAC Name 3-pyrrol-1-ylpropanenitrile
SMILES N#CCCN1C=CC=C1

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 120.15
Chemical Name or Material 3-(1H-Pyrrol-1-yl)propanenitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.