Learn More
3-(1H-Pyrrol-1-yl)propanenitrile, 97%, Maybridge™
Brand: Maybridge CD01949EA
Code : 31
Additional Details : CAS Number : 43036-06-2 Weight : 0.06000kg
Quantity | 10g |
---|
Chemical Identifiers
43036-06-2 | |
120.155 | |
3-1h-pyrrol-1-yl propanenitrile, n-2-cyanoethyl pyrrole, 1-2-cyanoethyl pyrrole, pyrrole-1-propionitrile, 1h-pyrrole-1-propanenitrile, 3-pyrrol-1-yl propiononitrile, n-2-cyanoethyl-pyrrole, 3-pyrrol-1-yl propanenitrile, n-cyanoethylpyrrole, n-2-cyanoethyl pyrole | |
3-pyrrol-1-ylpropanenitrile |
C7H8N2 | |
IYOLJLGYJMJLSU-UHFFFAOYSA-N | |
96527 | |
C1=CN(C=C1)CCC#N |
Specifications
3-(1H-Pyrrol-1-yl)propanenitrile | |
43036-06-2 | |
C7H8N2 | |
IYOLJLGYJMJLSU-UHFFFAOYSA-N | |
3-pyrrol-1-ylpropanenitrile | |
96527 |
10g | |
97% | |
3-1h-pyrrol-1-yl propanenitrile, n-2-cyanoethyl pyrrole, 1-2-cyanoethyl pyrrole, pyrrole-1-propionitrile, 1h-pyrrole-1-propanenitrile, 3-pyrrol-1-yl propiononitrile, n-2-cyanoethyl-pyrrole, 3-pyrrol-1-yl propanenitrile, n-cyanoethylpyrrole, n-2-cyanoethyl pyrole | |
C1=CN(C=C1)CCC#N | |
120.155 | |
120.15 |