Learn More
3-(1H-Pyrrol-1-yl)benzaldehyde, 95+%, Maybridge™
Brand: Maybridge CC21904DA
Code : 31
Additional Details : CAS Number : 129747-77-9 Weight : 0.05100kg
Quantity | 1g |
---|
Chemical Identifiers
129747-77-9 | |
171.199 | |
3-1h-pyrrol-1-yl benzaldehyde, 3-pyrrol-1-yl-benzaldehyde, 3-pyrrol-1-yl benzaldehyde, benzaldehyde,3-1h-pyrrol-1-yl, benzaldehyde, 3-1h-pyrrol-1-yl, acmc-20airb, 3-pyrrolylbenzaldehyde, 3-1-pyrrolyl benzaldehyde | |
3-pyrrol-1-ylbenzaldehyde |
C11H9NO | |
PALTUANHIBXQMX-UHFFFAOYSA-N | |
2776528 | |
C1=CN(C=C1)C2=CC=CC(=C2)C=O |
Specifications
3-(1H-Pyrrol-1-yl)benzaldehyde | |
1g | |
95% | |
3-1h-pyrrol-1-yl benzaldehyde, 3-pyrrol-1-yl-benzaldehyde, 3-pyrrol-1-yl benzaldehyde, benzaldehyde,3-1h-pyrrol-1-yl, benzaldehyde, 3-1h-pyrrol-1-yl, acmc-20airb, 3-pyrrolylbenzaldehyde, 3-1-pyrrolyl benzaldehyde | |
C1=CN(C=C1)C2=CC=CC(=C2)C=O | |
171.199 | |
171.2 |
Amber Glass Bottle | |
129747-77-9 | |
C11H9NO | |
PALTUANHIBXQMX-UHFFFAOYSA-N | |
3-pyrrol-1-ylbenzaldehyde | |
2776528 | |
95% |