missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Pyrazol-3-yl)benzoic acid, 97%, Thermo Scientific™

Product Code. 10694025
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10694025 5 g Amber Glass Bottle 5g
10053872 250 mg Amber Glass Bottle 250mg
10596903 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10694025 Supplier Thermo Scientific Maybridge Supplier No. CC31701DE
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 850375-11-0
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.19
MDL Number MFCD03407956
InChI Key RXZRBZWATRFHCS-UHFFFAOYSA-N
Synonym 3-1h-pyrazol-5-yl benzoic acid, 3-1h-pyrazol-3-yl benzoic acid, 3-2h-pyrazol-3-yl benzoic acid, benzoic acid, 3-1h-pyrazol-3-yl, 5-3-carboxyphenyl-1h-pyrazole, pubchem22748, buttpark 44\01-87, d0tx9p, 3-pyrazol-3-ylbenzoic acid, 3-pyrazol-5-ylbenzoic acid
PubChem CID 2795542
IUPAC Name 3-(1H-pyrazol-5-yl)benzoic acid
SMILES OC(=O)C1=CC=CC(=C1)C1=CC=NN1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 188.19
Percent Purity 97%
Chemical Name or Material 3-(1H-Pyrazol-3-yl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.