missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Pyrazol-1-ylmethyl)benzylamine, 97%, Thermo Scientific™

Product Code. 10275984
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10275984 250 mg 250mg
10356234 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10275984 Supplier Thermo Scientific Maybridge Supplier No. CC56113CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 562803-76-3
Molecular Formula C11H13N3
Molecular Weight (g/mol) 187.246
MDL Number MFCD08572164
InChI Key XKFHMAZBNKKNMT-UHFFFAOYSA-N
Synonym 3-1h-pyrazol-1-ylmethyl benzylamine, 3-1h-pyrazol-1-ylmethyl phenyl methanamine, 1-3-aminomethyl benzyl-1h-pyrazole, 3-pyrazol-1-ylmethyl phenyl methanamine, 1-3-pyrazol-1-ylmethyl phenyl methanamine, 3-pyrazolylmethyl phenyl methylamine, 3-1h-pyrazol-1-yl methyl benzylamine, 3-1h-pyrazole-1-ylmethyl benzenemethanamine, benzenemethanamine,3-1h-pyrazol-1-ylmethyl, 3-1h-pyrazol-1-yl methyl phenyl methanamine
PubChem CID 16640564
IUPAC Name [3-(pyrazol-1-ylmethyl)phenyl]methanamine
SMILES C1=CC(=CC(=C1)CN)CN2C=CC=N2

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 187.24
Percent Purity 90%
Chemical Name or Material 3-(1H-Pyrazol-1-ylmethyl)benzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.