missing translation for 'onlineSavingsMsg'
Learn More

[3-(1H-Pyrazol-1-yl)phenyl]methanol, ≥90%, Thermo Scientific™

Catalog Number p-3175976
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
864068-80-4
C10H10N2O
174.20
MFCD06740322
VINVOQJANISHSK-UHFFFAOYSA-N
3-1h-pyrazol-1-yl phenyl methanol, 3-1-pyrazolyl benzyl alcohol, 3-pyrazol-1-yl-phenyl-methanol, 3-pyrazol-1-yl phenyl methanol, 3-pyrazol-1-ylphenyl methanol, 3-pyrazolylphenyl methan-1-ol, 3-1h-pyrazole-1-yl benzyl alcohol, benzenemethanol, 3-1h-pyrazol-1-yl, benzenemethanol,3-1h-pyrazol-1-yl
7164580
[3-(1H-pyrazol-1-yl)phenyl]methanol
OCC1=CC(=CC=C1)N1C=CC=N1
This item is not returnable. View return policy
864068-80-4
C10H10N2O
174.20
MFCD06740322
VINVOQJANISHSK-UHFFFAOYSA-N
3-1h-pyrazol-1-yl phenyl methanol, 3-1-pyrazolyl benzyl alcohol, 3-pyrazol-1-yl-phenyl-methanol, 3-pyrazol-1-yl phenyl methanol, 3-pyrazol-1-ylphenyl methanol, 3-pyrazolylphenyl methan-1-ol, 3-1h-pyrazole-1-yl benzyl alcohol, benzenemethanol, 3-1h-pyrazol-1-yl, benzenemethanol,3-1h-pyrazol-1-yl
7164580
[3-(1H-pyrazol-1-yl)phenyl]methanol
OCC1=CC(=CC=C1)N1C=CC=N1
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 174.2
Percent Purity 90%
Chemical Name or Material [3-(1H-pyrazol-1-yl)phenyl]methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'