missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-indol-5-yl)propanoic acid, TRC

Product Code. 30400266
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400266 50 mg -
30405303 10 mg 10mg
30399786 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30400266 Supplier Toronto Research Chemicals Supplier No. TRCI62828850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 192717-19-4
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.21
IUPAC Name 3-(1H-indol-5-yl)propanoic acid
SMILES OC(=O)CCc1ccc2[nH]ccc2c1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C11H11NO2/c13-11(14)4-2-8-1-3-10-9(7-8)5-6-12-10/h1,3,5-7,12H,2,4H2,(H,13,14)
Formula Weight 189.079
Chemical Name or Material 3-(1H-indol-5-yl)propanoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.