missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Indol-2-yl)aniline, TRC

Product Code. 30390696
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390696 500 mg 500mg
30403421 100 mg -
30394999 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390696 Supplier Toronto Research Chemicals Supplier No. TRCI628310500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6318-72-5
Molecular Formula C14H12N2
Molecular Weight (g/mol) 208.25
Synonym 2-(3-Aminophenyl)-1H-indole, 2-(m-Aminophenyl)indole, NSC 31687
SMILES Nc1cccc(c1)c2cc3ccccc3[nH]2

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C14H12N2/c15-12-6-3-5-10(8-12)14-9-11-4-1-2-7-13(11)16-14/h1-9,16H,15H2
Chemical Name or Material 3-(1H-Indol-2-yl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.