missing translation for 'onlineSavingsMsg'
Learn More

3-(1h-1,2,4-Triazol-1-ylmethyl)benzaldehyde, 97%, Thermo Scientific™

Product Code. 10719215
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10719215 1 g 1g
10307483 250 mg 250mg
10123134 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10719215 Supplier Thermo Scientific Maybridge Supplier No. CC45404DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 876316-30-2
Molecular Formula C10H9N3O
Molecular Weight (g/mol) 187.202
InChI Key IBVURYHFXBPNCW-UHFFFAOYSA-N
Synonym 3-1h-1,2,4-triazol-1-ylmethyl benzaldehyde, benzaldehyde,3-1h-1,2,4-triazol-1-ylmethyl, 3-1,2,4-triazol-1-ylmethyl benzaldehyde, 3-1h-1,2,4-triazol-1-yl methyl benzaldehyde, 3-1,2,4-triazolylmethyl benzaldehyde
PubChem CID 22360585
IUPAC Name 3-(1,2,4-triazol-1-ylmethyl)benzaldehyde
SMILES C1=CC(=CC(=C1)CN2C=NC=N2)C=O

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 187.2
Percent Purity 97%
Chemical Name or Material 3-(1H-1, 2, 4-Triazol-1-ylmethyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.