missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(1h-1,2,4-Triazol-1-ylmethyl)benzaldehyde, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 187.2 |
Percent Purity | 97% |
Chemical Name or Material | 3-(1H-1, 2, 4-Triazol-1-ylmethyl)benzaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'