missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-1,2,4-triazol-1-ylmethyl)aniline, ≥97%, Thermo Scientific™

Product Code. 10205804
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10205804 1 g Amber Glass Bottle 1g
10518633 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10205804 Supplier Thermo Scientific Maybridge Supplier No. CC45414DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 127988-22-1
Molecular Formula C9H10N4
Molecular Weight (g/mol) 174.207
MDL Number MFCD08060528
InChI Key LFINNEVQJQXPHN-UHFFFAOYSA-N
Synonym 3-1h-1,2,4-triazol-1-ylmethyl aniline, 3-1,2,4-triazol-1-ylmethyl aniline, 3-1,2,4-triazol-1-yl methyl aniline, 3-1,2,4 triazol-1-ylmethyl-phenylamine, 3-1h-1,2,4-triazol-1-yl methyl aniline, benzenamine,3-1h-1,2,4-triazol-1-ylmethyl, 3-1,2,4-triazolylmethyl phenylamine, acmc-20e0ac, 1-3-aminobenzyl-1h-1,2,4-triazole
PubChem CID 6482004
IUPAC Name 3-(1,2,4-triazol-1-ylmethyl)aniline
SMILES C1=CC(=CC(=C1)N)CN2C=NC=N2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 174.21
Percent Purity 97%
Chemical Name or Material 3-(1H-1, 2, 4-Triazol-1-ylmethyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.