missing translation for 'onlineSavingsMsg'
Learn More

3-(1h-1,2,4-Triazol-1-yl)aniline, 90%, Thermo Scientific™

Product Code. 10653825
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 176032-78-3
Molecular Formula C8H8N4
Molecular Weight (g/mol) 160.18
InChI Key QOHJDIAWBVPZFL-UHFFFAOYSA-N
PubChem CID 23503656
IUPAC Name 3-(1,2,4-triazol-1-yl)aniline
SMILES C1=CC(=CC(=C1)N)N2C=NC=N2

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 3-1h-1,2,4-triazol-1-yl aniline, 3-1,2,4-triazol-1-yl aniline, benzenamine,3-1h-1,2,4-triazol-1-yl, benzenamine, 3-1h-1,2,4-triazol-1-yl, 1-3-aminophenyl-1h-1,2,4-triazole, 3-1,2,4-triazolyl phenylamine, 3-1,2,4 triazol-1-yl-phenylamine, 3-1,2,4 triazol-1-yl phenylamine
Formula Weight 160.18
Percent Purity 90%
Chemical Name or Material 3-(1H-1, 2, 4-Triazol-1-yl)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'