missing translation for 'onlineSavingsMsg'
Learn More

3,17-Di-O-acetyl Androsta-5,14,16-triene-3Beta,17-diol, TRC

Product Code. 30357749
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357749 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357749 Supplier Toronto Research Chemicals Supplier No. TRCD30868025MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,17-Di-O-acetyl Androsta-5,14,16-triene-3beta,17-diol
CAS 34635-42-2
Molecular Formula C23H30O4
Quantity 25 mg
Synonym Androsta-5,14,16-triene-3beta,17-diol, Diacetate
InChI Formula InChI=1S/C23H30O4/c1-14(24)26-17-9-11-22(3)16(13-17)5-6-18-19-7-8-21(27-15(2)25)23(19,4)12-10-20(18)22/h5,7-8,17-18,20H,6,9-13H2,1-4H3/t17-,18+,20+,22+,23+/m1/s1
SMILES CC(=O)O[C@@H]1CC[C@]2(C)[C@H]3CC[C@]4(C)C(=CC=C4[C@@H]3CC=C2C1)OC(=O)C
IUPAC Name [(3R,8R,9S,10R,13S)-17-acetyloxy-10,13-dimethyl-2,3,4,7,8,9,11,12-octahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate
Molecular Weight (g/mol) 370.48
Formula Weight 370.2144
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.