missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[3-(1-Pyrrolidinylmethyl)phenyl]methanamine, 97%, Thermo Scientific™
Specifications
Specifications
| CAS | 91271-78-2 |
| Assay Percent Range | 90% |
| Packaging | Amber Glass Bottle |
| Molecular Formula | C12H18N2 |
| MDL Number | MFCD06408780 |
| Quantity | 250mg |
| Synonym | 3-pyrrolidin-1-ylmethyl-benzylamine, 1-3-pyrrolidin-1-ylmethyl phenyl methanamine, 3-1-pyrrolidinylmethyl phenyl methanamine, 3-pyrrolidin-1-ylmethyl phenyl methanamine, 3-pyrrolidin-1-ylmethylbenzylamine, 3-pyrrolidinylmethyl phenyl methylamine, 3-pyrrolidin-1-ylmethylphenylamine, 3-1-pyrrolidinyl methyl benzylamine, 3-1-pyrrolidinylmethyl phenyl methanamine-7 |
| InChI Key | RTUWXPVILJUANC-UHFFFAOYSA-N |
| SMILES | NCC1=CC=CC(CN2CCCC2)=C1 |
| IUPAC Name | [3-(pyrrolidin-1-ylmethyl)phenyl]methanamine |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?