missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Piperazinyl)-1,2-benzisoxazole, 96%, Thermo Scientific™

Product Code. 11464680
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11464680 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11464680 Supplier Thermo Scientific Alfa Aesar Supplier No. H50746.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87691-89-2
Molecular Formula C11H13N3O
Molecular Weight (g/mol) 203.245
MDL Number MFCD08060968
InChI Key ZDFQBFVFCPABKQ-UHFFFAOYSA-N
Synonym 3-piperazin-1-yl benzo d isoxazole, 3-1-piperazinyl-1,2-benzisoxazole, 3-piperazin-1-yl-1,2-benzoxazole, 3-piperazin-1-yl-1,2-benzisoxazole, 3-1-piperazino-1,2-benzisoxazole, 3-piperazinylbenzo d isoxazole, 3-piperazin-1-yl-benzo d isoxazole, 3-1-piperazinyl-1,2-benzoxazole, 4 1,2-benzisoxazol-3-yl piperazine
PubChem CID 13232475
IUPAC Name 3-piperazin-1-yl-1,2-benzoxazole
SMILES C1CN(CCN1)C2=NOC3=CC=CC=C32

Specifications

Melting Point 74°C to 76°C
Quantity 1 g
Formula Weight 203.25
Percent Purity 96%
Chemical Name or Material 3-(1-Piperazinyl)-1,2-benzisoxazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.