missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Naphthyl)-L-alanine, 95%

Product Code. 15400359
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15400359 1 g 1g
15420359 250 mg 250mg
15410359 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15400359 Supplier Thermo Scientific Alfa Aesar Supplier No. H63586.03
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 55516-54-6 | C13H13NO2 | 215.25 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55516-54-6
Molecular Formula C13H13NO2
Molecular Weight (g/mol) 215.25
MDL Number MFCD00066086
InChI Key OFYAYGJCPXRNBL-LBPRGKRZSA-N
Synonym l-1-naphthylalanine, 3-1-naphthyl-l-alanine, h-1-nal-oh, s-2-amino-3-naphthalen-1-yl propanoic acid, 3-1-naphthyl-l-ala, unii-9g930ul92r, naphthalen-2-yl-3-alanine, 2s-2-amino-3-naphthalen-1-yl propanoic acid, 3-naphth-1-yl-d-alanine, 1-naphthyl-l-alanine
PubChem CID 2724883
SMILES [NH3+][C@@H](CC1=C2C=CC=CC2=CC=C1)C([O-])=O

Specifications

Beilstein 4352833
Solubility Information Slightly soluble in water.
IUPAC Name (2S)-2-azaniumyl-3-(naphthalen-1-yl)propanoate
Quantity 1 g
Formula Weight 215.25
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-(1-Naphthyl)-L-alanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.