missing translation for 'onlineSavingsMsg'
Learn More

n-Hexyl n-octyl ether, 98%, Thermo Scientific™

Product Code. 11438967
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11438967 5 g 5g
11448967 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11438967 Supplier Thermo Scientific Alfa Aesar Supplier No. B20576.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17071-54-4
Molecular Formula C14H30O
Molecular Weight (g/mol) 214.393
MDL Number MFCD00049132
InChI Key KUTWKGLRMXBROO-UHFFFAOYSA-N
Synonym hexyl octyl ether, n-hexyl n-octyl ether, 1-hexyloxy octane, octylhexyl ether, 1-n-hexyloxyoctane, 1-hexyloxy octane #
PubChem CID 519362
IUPAC Name 1-hexoxyoctane
SMILES CCCCCCCCOCCCCCC

Specifications

Quantity 5 g
Formula Weight 214.39
Percent Purity 98%
Chemical Name or Material n-Hexyl n-octyl ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.