missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Methyl-1H-pyrazol-5-yl)benzylamine, 97%, Thermo Scientific™

Product Code. 10081974
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10081974 1 g Amber Glass Bottle 1g
10618814 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10081974 Supplier Thermo Scientific Maybridge Supplier No. CC60513DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 934570-45-3
Molecular Formula C11H13N3
Molecular Weight (g/mol) 187.246
MDL Number MFCD09702399
InChI Key UPQOTZRZQXMJNT-UHFFFAOYSA-N
Synonym 3-1-methyl-1h-pyrazol-5-yl phenyl methanamine, 3-1-methyl-1h-pyrazol-5-yl benzylamine, 3-2-methylpyrazol-3-yl phenyl methanamine, 3-1-methylpyrazol-5-yl phenyl methylamine, 3-1-methyl-1h-pyrazol-5-yl phenyl methylamine, 1-3-1-methyl-1h-pyrazol-5-yl phenyl methanamine, 1-3-2-methylpyrazol-3-yl phenyl methanamine, benzenemethanamine,3-1-methyl-1h-pyrazol-5-yl
PubChem CID 24229625
IUPAC Name [3-(2-methylpyrazol-3-yl)phenyl]methanamine
SMILES CN1C(=CC=N1)C2=CC(=CC=C2)CN

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 187.24
Percent Purity 97%
Chemical Name or Material 3-(1-Methyl-1H-pyrazol-5-yl)benzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.