missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Methyl-1H-pyrazol-5-yl)benzoic acid, 97%, Thermo Scientific™

Product Code. 10708275
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10708275 250 mg Amber Glass Bottle 250mg
10613635 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10708275 Supplier Thermo Scientific Maybridge Supplier No. CC60501CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 628297-55-2
Molecular Formula C11H10N2O2
Molecular Weight (g/mol) 202.213
MDL Number MFCD09702398
InChI Key OBOMYSVFLSMYLV-UHFFFAOYSA-N
Synonym 3-1-methyl-1h-pyrazol-5-yl benzoic acid, 3-2-methylpyrazol-3-yl benzoic acid, 3-1-methylpyrazol-5-yl benzoic acid, benzoicacid,3-1-methyl-1h-pyrazol-5-yl, benzoicacid, 3-1-methyl-1h-pyrazol-5-yl, benzoic acid, 3-1-methyl-1h-pyrazol-5-yl
PubChem CID 18548847
IUPAC Name 3-(2-methylpyrazol-3-yl)benzoic acid
SMILES CN1C(=CC=N1)C2=CC(=CC=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 202.21
Percent Purity 97%
Chemical Name or Material 3-(1-Methyl-1H-pyrazol-5-yl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.