missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Methyl-1H-pyrazol-3-yl)benzonitrile, 95%, Thermo Scientific™

Product Code. 10453014
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10453014 1 g Amber Glass Bottle 1g
10738655 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10453014 Supplier Thermo Scientific Maybridge Supplier No. CC60616DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 207909-05-5
Molecular Formula C11H9N3
Molecular Weight (g/mol) 183.214
MDL Number MFCD09065009
InChI Key FLALRGXSUSNEAF-UHFFFAOYSA-N
Synonym 3-1-methyl-1h-pyrazol-3-yl benzonitrile, 3-1-methylpyrazol-3-yl benzonitrile, benzonitrile, 3-1-methyl-1h-pyrazol-3-yl, 3-1-methylpyrazol-3-yl benzenecarbonitrile
PubChem CID 10655058
IUPAC Name 3-(1-methylpyrazol-3-yl)benzonitrile
SMILES CN1C=CC(=N1)C2=CC=CC(=C2)C#N

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 183.21
Percent Purity 95%
Chemical Name or Material 3-(1-Methyl-1H-pyrazol-3-yl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.