missing translation for 'onlineSavingsMsg'
Learn More

3-(1-methyl-1H-pyrazol-3-yl)aniline, 97+%, Thermo Scientific™

Product Code. 10536425
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10536425 5 g Amber Glass Bottle 5g
10161583 250 mg Amber Glass Bottle 250mg
10317264 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10536425 Supplier Thermo Scientific Maybridge Supplier No. CC60614DE
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 175202-37-6
Molecular Formula C10H11N3
Molecular Weight (g/mol) 173.219
InChI Key ZRXOZSROZJGRRH-UHFFFAOYSA-N
Synonym 3-1-methyl-1h-pyrazol-3-yl aniline, 3-1-methylpyrazol-3-yl aniline, 3-1-methyl-1h-pyrazol-3-yl-phenylamine, benzenamine, 3-1-methyl-1h-pyrazol-3-yl, 3-1-methylpyrazol-3-yl phenylamine, 3-1-methyl-1h-pyrazol-3-yl phenylamine, 3-3-aminophenyl-1-methyl-1h-pyrazole
PubChem CID 2735396
IUPAC Name 3-(1-methylpyrazol-3-yl)aniline
SMILES CN1C=CC(=N1)C2=CC(=CC=C2)N

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 173.22
Percent Purity 97%
Chemical Name or Material 3-(1-Methyl-1H-pyrazol-3-yl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.