missing translation for 'onlineSavingsMsg'
Learn More

3-(1-methyl-1H-pyrazol-3-yl)aniline, 97+%, Thermo Scientific™

Catalog Number 10161583
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
175202-37-6
C10H11N3
173.219
ZRXOZSROZJGRRH-UHFFFAOYSA-N
3-1-methyl-1h-pyrazol-3-yl aniline, 3-1-methylpyrazol-3-yl aniline, 3-1-methyl-1h-pyrazol-3-yl-phenylamine, benzenamine, 3-1-methyl-1h-pyrazol-3-yl, 3-1-methylpyrazol-3-yl phenylamine, 3-1-methyl-1h-pyrazol-3-yl phenylamine, 3-3-aminophenyl-1-methyl-1h-pyrazole
2735396
3-(1-methylpyrazol-3-yl)aniline
CN1C=CC(=N1)C2=CC(=CC=C2)N
This item is not returnable. View return policy
175202-37-6
C10H11N3
173.219
ZRXOZSROZJGRRH-UHFFFAOYSA-N
3-1-methyl-1h-pyrazol-3-yl aniline, 3-1-methylpyrazol-3-yl aniline, 3-1-methyl-1h-pyrazol-3-yl-phenylamine, benzenamine, 3-1-methyl-1h-pyrazol-3-yl, 3-1-methylpyrazol-3-yl phenylamine, 3-1-methyl-1h-pyrazol-3-yl phenylamine, 3-3-aminophenyl-1-methyl-1h-pyrazole
2735396
3-(1-methylpyrazol-3-yl)aniline
CN1C=CC(=N1)C2=CC(=CC=C2)N
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 173.22
Percent Purity 97%
Chemical Name or Material 3-(1-Methyl-1H-pyrazol-3-yl)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'