missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Azepanyl)-2,2-dimethylpropylamine, ≥95%, Thermo Scientific™

Product Code. 10503983
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10503983

Brand: Thermo Scientific Maybridge MO01190DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 845885-85-0
Molecular Formula C11H24N2
Molecular Weight (g/mol) 184.327
MDL Number MFCD06200874
InChI Key SUDFIJJDSKVMSO-UHFFFAOYSA-N
Synonym 3-azepan-1-yl-2,2-dimethylpropan-1-amine, 3-1-azepanyl-2,2-dimethylpropylamine, 3-azepan-1-yl-2,2-dimethyl-propylamine, 1h-azepine-1-propanamine,hexahydro-b,b-dimethyl, 3-azaperhydroepinyl-2,2-dimethylpropylamine
PubChem CID 2794705
IUPAC Name 3-(azepan-1-yl)-2,2-dimethylpropan-1-amine
SMILES CC(C)(CN)CN1CCCCCC1

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 184.32
Percent Purity 95%
Chemical Name or Material 3-(1-Azepanyl)-2, 2-dimethylpropylamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.