missing translation for 'onlineSavingsMsg'
Learn More

3-(1,4-Dihydro-5-methyl-4-oxo-7-propylimidazo[5,1-f][1,2,4]triazin-2-yl)-4-ethoxybenzenesulfonic Acid, TRC

Product Code. 30386141
Change view
Click to view available options
Quantity:
100 mg
1 g
2.5 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386141 100 mg -
30373955 2.5 g 2.5g
30373653 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30386141 Supplier Toronto Research Chemicals Supplier No. TRCD453410100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 437717-43-6
Molecular Formula C17 H20 N4 O5 S
Molecular Weight (g/mol) 392.43
Synonym Benzenesulfonic acid, 3-(1,4-dihydro-5-methyl-4-oxo-7-propylimidazo[5,1-f][1,2,4]triazin-2-yl)-4-ethoxy-, 3-(1,4-Dihydro-5-methyl-4-oxo-7-propylimidazo[5,1-f][1,2,4]triazin-2-yl)-4-ethoxybenzenesulfonic acid, 4-Ethoxy-3-(5-methyl-4-oxo-7-propyl-3,4-dihydroimidazo[5,1-f][1,2,4]triazin-2-yl)benzenesulfonic acid, Vardenafil Desethylpiperazinyl Sulfonic Acid
IUPAC Name 4-ethoxy-3-(5-methyl-4-oxo-7-propyl-3H-imidazo[5,1-f][1,2,4]triazin-2-yl)benzenesulfonic acid
SMILES CCCc1nc(C)c2C(=O)NC(=Nn12)c3cc(ccc3OCC)S(=O)(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C17H20N4O5S/c1-4-6-14-18-10(3)15-17(22)19-16(20-21(14)15)12-9-11(27(23,24)25)7-8-13(12)26-5-2/h7-9H,4-6H2,1-3H3,(H,19,20,22)(H,23,24,25)
Formula Weight 392.1154
Chemical Name or Material Vardenafil Desethylpiperazinyl sulfonic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.