missing translation for 'onlineSavingsMsg'
Learn More

[3-(1,3-Thiazol-2-yl)phenyl]methylamine, 97%, Thermo Scientific™

Product Code. 10406693
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10406693 1 g Amber Glass Bottle 1g
10022274 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10406693 Supplier Thermo Scientific Maybridge Supplier No. CC36513DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 672324-88-8
Molecular Formula C10H10N2S
Molecular Weight (g/mol) 190.264
MDL Number MFCD08741424
InChI Key UGCRMJJXRCJZHJ-UHFFFAOYSA-N
Synonym 3-1,3-thiazol-2-yl phenyl methylamine, 3-1,3-thiazol-2-yl phenyl methanamine, 1-3-1,3-thiazol-2-yl phenyl methanamine, 3-thiazol-2-yl phenyl methanamine, 3-thiazol-2-yl benzylamine, benzenemethanamine, 3-2-thiazolyl, 2-3-aminomethyl phenyl-1,3-thiazole, benzenemethanamine,3-2-thiazolyl
PubChem CID 18525727
IUPAC Name [3-(1,3-thiazol-2-yl)phenyl]methanamine
SMILES C1=CC(=CC(=C1)CN)C2=NC=CS2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 190.27
Percent Purity 97%
Chemical Name or Material [3-(1, 3-Thiazol-2-yl)phenyl]methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.