Learn More
[3-(1,3-Thiazol-2-yl)phenyl]methylamine, 97%, Maybridge
Brand: Maybridge CC36513CB
Code : 31
Additional Details : CAS Number : 672324-88-8 Weight : 0.05025kg Transport : UN number : 2735 Chem class : 8 Pack group : III
Packaging | Amber Glass Bottle |
---|---|
Quantity | 250mg |
Chemical Identifiers
672324-88-8 | |
190.264 | |
UGCRMJJXRCJZHJ-UHFFFAOYSA-N | |
18525727 | |
C1=CC(=CC(=C1)CN)C2=NC=CS2 |
C10H10N2S | |
MFCD08741424 | |
3-1,3-thiazol-2-yl phenyl methylamine, 3-1,3-thiazol-2-yl phenyl methanamine, 1-3-1,3-thiazol-2-yl phenyl methanamine, 3-thiazol-2-yl phenyl methanamine, 3-thiazol-2-yl benzylamine, benzenemethanamine, 3-2-thiazolyl, 2-3-aminomethyl phenyl-1,3-thiazole, benzenemethanamine,3-2-thiazolyl | |
[3-(1,3-thiazol-2-yl)phenyl]methanamine |
Specifications
250mg | |
Amber Glass Bottle | |
96.5 | |
95% | |
MFCD08741424 | |
UGCRMJJXRCJZHJ-UHFFFAOYSA-N | |
[3-(1,3-thiazol-2-yl)phenyl]methanamine | |
18525727 | |
97% |
[3-(1, 3-Thiazol-2-yl)phenyl]methylamine | |
672324-88-8 | |
100.0 | |
C10H10N2S | |
3-1,3-thiazol-2-yl phenyl methylamine, 3-1,3-thiazol-2-yl phenyl methanamine, 1-3-1,3-thiazol-2-yl phenyl methanamine, 3-thiazol-2-yl phenyl methanamine, 3-thiazol-2-yl benzylamine, benzenemethanamine, 3-2-thiazolyl, 2-3-aminomethyl phenyl-1,3-thiazole, benzenemethanamine,3-2-thiazolyl | |
C1=CC(=CC(=C1)CN)C2=NC=CS2 | |
190.264 | |
190.27 |