missing translation for 'onlineSavingsMsg'
Learn More

[3-(1,3-Thiazol-2-yl)phenyl]methanol, 97%, Thermo Scientific™

Product Code. 10559793
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 184847-97-0
Molecular Formula C10H9NOS
Molecular Weight (g/mol) 191.248
MDL Number MFCD09025828
InChI Key VDFDKGHCZKGOLZ-UHFFFAOYSA-N
Synonym 3-1,3-thiazol-2-yl phenyl methanol, 3-thiazol-2-yl benzyl alcohol, 3-thiazol-2-yl phenyl methanol, 3-1,3-thiazol-2-yl phenyl methan-1-ol
PubChem CID 18525726
IUPAC Name [3-(1,3-thiazol-2-yl)phenyl]methanol
SMILES C1=CC(=CC(=C1)CO)C2=NC=CS2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 191.25
Percent Purity 97%
Chemical Name or Material [3-(1, 3-Thiazol-2-yl)phenyl]methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'