missing translation for 'onlineSavingsMsg'
Learn More

3-(1,3-Benzothiazol-2-yl)aniline, Thermo Scientific™

Product Code. 10763137
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10763137 5 g Amber Glass Bottle 5g
10619705 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10763137 Supplier Thermo Scientific Maybridge Supplier No. MO08615DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 41230-21-1
Molecular Formula C13H10N2S
Molecular Weight (g/mol) 226.297
InChI Key MKMRZZJJDIBAJF-UHFFFAOYSA-N
Synonym 3-benzothiazol-2-yl-phenylamine, 3-1,3-benzothiazol-2-yl aniline, 3-benzothiazol-2-ylphenylamine, benzenamine,3-2-benzothiazolyl, cambridge id 5250055, 3-2-benzothiazolyl aniline, 3-benzothiazol-2-yl aniline, 3-benzo d thiazol-2-yl aniline, 3-benzothiazol-2-yl benzenamine, 3-1,3-benzothiazol-2-yl phenyl amine
PubChem CID 720999
IUPAC Name 3-(1,3-benzothiazol-2-yl)aniline
SMILES C1=CC=C2C(=C1)N=C(S2)C3=CC(=CC=C3)N

Specifications

Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 226.3
Percent Purity 97%
Chemical Name or Material 3-(1, 3-benzothiazol-2-yl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.