missing translation for 'onlineSavingsMsg'
Learn More

3-(1,3-Benzothiazol-2-yl)-4-methylthiophen-2-amine, Thermo Scientific™

Product Code. 10427184
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10427184 5 g 5g
10563202 1 g 1g
10600106 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10427184 Supplier Thermo Scientific Maybridge Supplier No. GK03755DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 232941-00-3
Molecular Formula C12H10N2S2
Molecular Weight (g/mol) 246.35
MDL Number MFCD02179980
InChI Key YXXFAQKMMOVVIA-UHFFFAOYSA-N
Synonym 3-1,3-benzothiazol-2-yl-4-methylthiophen-2-amine, 2-2-amino-4-methylthien-3-yl benzothiazole, 3-benzo d thiazol-2-yl-4-methylthiophen-2-amine, 2-thiophenamine,3-2-benzothiazolyl-4-methyl, maybridge1_003812, 3-benzothiazol-2-yl-4-methyl-2-thienylamine, 2-2-amino-4-methylthien-3-yl-1,3-benzothiazol, 2-2-amino-4-methylthien-3-yl-1,3-benzothiazole, 3-1,3-benzothiazol-2-yl-4-methyl-2-thiophenamine, 3-1,3-benzothiazol-2-yl-4-methyl-thiophen-2-amine
PubChem CID 2735376
IUPAC Name 3-(1,3-benzothiazol-2-yl)-4-methylthiophen-2-amine
SMILES CC1=CSC(N)=C1C1=NC2=CC=CC=C2S1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 246.36
Percent Purity 97%
Chemical Name or Material 3-(1, 3-Benzothiazol-2-yl)-4-methylthiophen-2-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.