missing translation for 'onlineSavingsMsg'
Learn More

(2S,5R,6R)-6-Amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid (6-Aminopenicillanic Acid), Mikromol™

Product Code. 30052449
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052449 100 mg 100mg
30050125 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30052449 Supplier Mikromol Supplier No. MM0026.04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2S,5R,6R)-6-Amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid (6-Aminopenicillanic Acid)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-Aminopenicillanic Acid
CAS 551-16-6
Analyte or Component Names 6-Aminopenicillanic Acid
Shipping Condition Ice pack (-20°C)
Molecular Formula C8 H12 N2 O3 S
Certifications/Compliance ISO 17025
Synonym Amoxicillin Trihydrate Imp. A (EP), Amoxicillin Imp. A (EP), Ampicillin Sodium Imp. A (EP), Ampicillin Imp. A (EP), Benzylpenicillin Potassium Imp. A (EP), Benzylpenicillin Imp. A (EP), Cloxacillin Sodium Imp. C (EP), Cloxacillin Imp. C (EP), Dicloxacillin Sodium Imp. C (EP), Dicloxacillin Imp. C (EP), Flucloxacillin Sodium Imp. C (EP), Flucloxacillin Imp. C (EP), Oxacillin Sodium Imp. A (EP), Oxacillin Imp. A (EP), Phenoxymethylpenicillin Potassium Imp. C (EP), Phenoxymethylpenicillin Imp. C (EP), Piperacillin Sodium Imp. H (EP), Piperacillin Imp. H (EP), Amoxicillin USP Related Compound A, Amoxicillin USP RC A, 6-Aminopenicillanic Acid, (2S,5R,6R)-6-Amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid, 6-APA, BRL-955, USP Amoxicillin Related Compound A, Ph Eur Amoxicillin Impurity A, Penicinic acid (1), Penicinic acid (2), 8-Hydroxypenillic acid (8-HPA), 6-APA, 6-Aminopenicillanic acid dimer (6-APA dimer), Amoxicillin Related Compound A, Amoxicillin Trihydrate Impurity A, Ampicillin Sodium Impurity A, Benzylpenicillin Potassium Impurity A, Carbenicillin Sodium Impurity A, Cloxacillin Sodium Impurity C, Dicloxacillin Sodium Impurity C, Flucloxacillin Sodium Impurity C, Oxacillin Sodium Monohydrate Impurity A, Phenoxymethylpenicillin Potassium Impurity C, Piperacillin Sodium Impurity H, Sulbactam Sodium Impurity B, Amoxicillin Impurity A, Ampicillin Impurity A, Benzylpenicillin Impurity A, Carbenicillin Impurity A, Cloxacillin Impurity C, Dicloxacillin Impurity C, Flucloxacillin Impurity C, Oxacillin Impurity A, Phenoxymethylpenicillin Impurity C, Piperacillin Impurity H, Sulbactam Impurity B
InChI Formula InChI=1S/C8H12N2O3S/c1-8(2)4(7(12)13)10-5(11)3(9)6(10)14-8/h3-4,6H,9H2,1-2H3,(H,12,13)/t3-,4+,6-/m1/s1
SMILES CC1(C)S[C@@H]2[C@H](N)C(=O)N2[C@H]1C(=O)O
IUPAC Name (2S,5R,6R)-6-amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Molecular Weight (g/mol) 216.26
Quantity 100 mg
Formula Weight 216.0569 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.