missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2S,4S)-2,4-Bis(diphenylphosphino)pentane, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | (2S,4S)-2,4-Bis(diphenylphosphino)pentane |
| CAS | 77876-39-2 |
| Molecular Formula | C29H30P2 |
| Quantity | 100 mg |
| Synonym | (2S,4S)-Pentane-2,4-diylbis(diphenylphosphine), 1,1'-[(1S,3S)-1,3-Dimethyl-1,3-propanediyl]bis[1,1-diphenylphosphine], (-)-2,4-BDPP |
| InChI Formula | InChI=1S/C29H30P2/c1-24(30(26-15-7-3-8-16-26)27-17-9-4-10-18-27)23-25(2)31(28-19-11-5-12-20-28)29-21-13-6-14-22-29/h3-22,24-25H,23H2,1-2H3/t24-,25-/m0/s1 |
| SMILES | C[C@@H](C[C@H](C)P(c1ccccc1)c2ccccc2)P(c3ccccc3)c4ccccc4 |
| IUPAC Name | [(2S,4S)-4-diphenylphosphanylpentan-2-yl]-diphenylphosphane |
| Molecular Weight (g/mol) | 440.5 |
| Formula Weight | 440.1823 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction