missing translation for 'onlineSavingsMsg'
Learn More

(2S,4R)-4-Hydroxy-L-glutamic Acid, TRC

Product Code. 30388825
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388825 50 mg -
30389833 10 mg -
30404378 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30388825 Supplier Toronto Research Chemicals Supplier No. TRCH94255050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2485-33-8
Molecular Formula C5H9NO5
Molecular Weight (g/mol) 163.13
Synonym (4R)-4-Hydroxy-L-glutamic Acid, (2S,4R)-g-4-Hydroxy-L-glutamic Acid, L-erythro-4-Hydroxyglutamic Acid, erythro-4-Hydroxy-L-glutamic Acid, H-(2S,4R)-Glu(g-OH)-OH
IUPAC Name (2S,4R)-2-amino-4-hydroxypentanedioic acid
SMILES N[C@@H](C[C@@H](O)C(=O)O)C(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C5H9NO5/c6-2(4(8)9)1-3(7)5(10)11/h2-3,7H,1,6H2,(H,8,9)(H,10,11)/t2-,3+/m0/s1
Formula Weight 163.0481
Chemical Name or Material (2S,4R)-4-Hydroxy-L-glutamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.