missing translation for 'onlineSavingsMsg'
Learn More

(2S,4R)-4-Hydroxy-1-[(4-nitrophenyl)methoxycarbonyl]pyrrolidine-2-carboxylic Acid, Mikromol

Product Code. 30463204
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30463204 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30463204 Supplier Mikromol Supplier No. MM1327.040025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2S,4R)-4-Hydroxy-1-[(4-nitrophenyl)methoxycarbonyl]pyrrolidine-2-carboxylic Acid, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2S,4R)-4-Hydroxy-1,2-pyrrolidinedicarboxylic Acid 1-(4-Nitrobenzyl) Ester
CAS 96034-57-0
Analyte or Component Names (2S,4R)-4-Hydroxy-1,2-pyrrolidinedicarboxylic Acid 1-(4-Nitrobenzyl) Ester
Shipping Condition Ice pack (-20°C)
Molecular Formula C13H14N2O7
Certifications/Compliance ISO 9001
Synonym (2S-trans)-4-Hydroxy-1,2-pyrrolidinedicarboxylic Acid 1-[(4-Nitrophenyl)methyl] Ester, (2S,4R)-1-(p-Nitrobenzyloxycarbonyl)-4-hydroxyproline, trans-4-Hydroxy-1-(4-nitrobenzyloxycarbonyl)-L-proline;
Solution Type Neat
SMILES O[C@@H]1C[C@H](N(C1)C(=O)OCc2ccc(cc2)[N+](=O)[O-])C(=O)O
Molecular Weight (g/mol) 310.26
Quantity 25 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.