missing translation for 'onlineSavingsMsg'
Learn More

(2S,4R)-4-Hydroxy-1,2-pyrrolidinedicarboxylic Acid 1-(4-Nitrobenzyl) Ester, TRC

Product Code. 30401618
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30401618 10 g
30393824 100 g
30392942 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30401618 Supplier Toronto Research Chemicals Supplier No. TRCH95321510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96034-57-0
Molecular Formula C13H14N2O7
Molecular Weight (g/mol) 310.26
Synonym 1,2-Pyrrolidinedicarboxylic acid, 4-hydroxy-, 1-[(4-nitrophenyl)methyl] ester, (2S,4R)-, 1,2-Pyrrolidinedicarboxylic acid, 4-hydroxy-, 1-[(4-nitrophenyl)methyl] ester, (2S-trans)-, (2S,4R)-1-(4-Nitrobenzyloxycarbonyl)-4-hydroxypyrrolidine-2-carboxylic acid, (2S,4R)-1-(p-Nitrobenzyloxycarbonyl)-4-hydroxyproline, trans-4-Hydroxy-1-(4-nitrobenzyloxycarbonyl)-L-proline, (2S-trans)-4-Hydroxy-1,2-pyrrolidinedicarboxylic Acid 1-[(4-Nitrophenyl)methyl] Ester, (2S,4R)-1-(p-Nitrobenzyloxycarbonyl)-4-hydroxyproline, trans-4-Hydroxy-1-(4-nitrobenzyloxycarbonyl)-L-proline
IUPAC Name (2S,4R)-4-hydroxy-1-[(4-nitrophenyl)methoxycarbonyl]pyrrolidine-2-carboxylic acid
SMILES O[C@@H]1C[C@H](N(C1)C(=O)OCc2ccc(cc2)[N+](=O)[O-])C(=O)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C13H14N2O7/c16-10-5-11(12(17)18)14(6-10)13(19)22-7-8-1-3-9(4-2-8)15(20)21/h1-4,10-11,16H,5-7H2,(H,17,18)/t10-,11+/m1/s1
Formula Weight 310.0801
Chemical Name or Material (2S,4R)-4-Hydroxy-1,2-pyrrolidinedicarboxylic Acid 1-(4-Nitrobenzyl) Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.