missing translation for 'onlineSavingsMsg'
Learn More

(2S,4R)-4-Ethyl-L-proline Hydrochloride, TRC

Product Code. 30386692
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30386692 10 mg
30376380 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30386692 Supplier Toronto Research Chemicals Supplier No. TRCE92565010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C7 H13 N O2 . H Cl
Molecular Weight (g/mol) 179.645
Synonym (2S-trans)-4-Ethyl proline Hydrochloride
IUPAC Name (2S,4R)-4-ethylpyrrolidine-2-carboxylic acid;hydrochloride
SMILES Cl.CC[C@H]1CN[C@@H](C1)C(=O)O

Specifications

InChI Formula InChI=1S/C7H13NO2.ClH/c1-2-5-3-6(7(9)10)8-4-5;/h5-6,8H,2-4H2,1H3,(H,9,10);1H/t5-,6+;/m1./s1
Quantity 10 mg
Formula Weight 179.071
Shipping Condition Room Temperature
Chemical Name or Material (2S,4R)-4-Ethyl-L-proline Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.