missing translation for 'onlineSavingsMsg'
Learn More

(2S,4E,6R,8S)-8-Hydroxy-2,4,6-trimethyl-4-nonenoic Acid Ethyl Ester, TRC

Product Code. 30397350
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397350 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30397350 Supplier Toronto Research Chemicals Supplier No. TRCH9723555MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2S,4E,6R,8S)-8-Hydroxy-2,4,6-trimethyl-4-nonenoic Acid Ethyl Ester
CAS 943858-34-2
Molecular Formula C14H26O3
Quantity 5 mg
InChI Formula InChI=1S/C14H26O3/c1-6-17-14(16)12(4)8-10(2)7-11(3)9-13(5)15/h7,11-13,15H,6,8-9H2,1-5H3/b10-7+/t11-,12-,13-/m0/s1
SMILES CCOC(=O)[C@@H](C)C\C(=C\[C@H](C)C[C@H](C)O)\C
IUPAC Name ethyl (E,2S,6R,8S)-8-hydroxy-2,4,6-trimethylnon-4-enoate
Molecular Weight (g/mol) 242.35
Formula Weight 242.1882
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.