missing translation for 'onlineSavingsMsg'
Learn More

(2S,3S,4S,5R,6R)-3,4,5-Trihydroxy-6-(6-methyl-1,3,4,14b-tetrahydrobenzo[c]pyrazino[1,2-a]pyrido[3,2-f]azepin-6-ium-2(10H)-yl)tetrahydro-2H-pyran-2-carboxylate, TRC

Product Code. 30457268
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457268 2.5 mg 2.5mg
30453948 5 mg 5mg
30450828 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30457268 Supplier Toronto Research Chemicals Supplier No. TRCT7949202.5MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2S,3S,4S,5R,6R)-3,4,5-Trihydroxy-6-(6-methyl-1,3,4,14b-tetrahydrobenzo[c]pyrazino[1,2-a]pyrido[3,2-f]azepin-6-ium-2(10H)-yl)tetrahydro-2H-pyran-2-carboxylate
Analyte or Component Names (2S,3S,4S,5R,6R)-3,4,5-Trihydroxy-6-(6-methyl-1,3,4,14b-tetrahydrobenzo[c]pyrazino[1,2-a]pyrido[3,2-f]azepin-6-ium-2(10H)-yl)tetrahydro-2H-pyran-2-carboxylate
Shipping Condition Room Temperature
Molecular Formula C23 H20 F N O5
InChI Formula InChI=1S/C23H20FNO5/c1-29-19-8-4-15-5-9-20(11-17(15)10-19)30-14-23(22(27)28)12-25(13-23)21(26)16-2-6-18(24)7-3-16/h2-11H,12-14H2,1H3,(H,27,28)
Solution Type Neat
SMILES COc1ccc2ccc(OCC3(CN(C3)C(=O)c4ccc(F)cc4)C(=O)O)cc2c1
IUPAC Name 1-(4-fluorobenzoyl)-3-[(7-methoxynaphthalen-2-yl)oxymethyl]azetidine-3-carboxylic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 409.407
Quantity 2.5 mg
Formula Weight 409.133
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.