Learn More
(2S,3S)-3-(N-BOC-amino)-1-oxirane-4-phenylbutane, 98%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 98737-29-2 |
---|---|
Molecular Formula | C15H21NO3 |
Molecular Weight (g/mol) | 263.34 |
MDL Number | MFCD02258997 |
InChI Key | NVPOUMXZERMIJK-QWHCGFSZSA-N |
Synonym | 2s,3s-1,2-epoxy-3-boc-amino-4-phenylbutane, 2s,3s-n-t-boc-3-amino-1,2-epoxy-4-phenylbutane, tert-butyl s-1-s-oxiran-2-yl-2-phenylethyl carbamate, 2s,3s-1,2-epoxy-3-tert-butoxycarbonylamino-4-phenylbutane, tert-butyl n-1s-1-2s-oxiran-2-yl-2-phenylethyl carbamate, tert-butyl s-r*,r*---1-oxiranyl-2-phenylethyl carbamate, tert-butyl n-1s-1-2s-oxiran-2-yl-2-phenyl-ethyl carbamate, 2s,3s---3-t-boc-amino-1,2-epoxy-4-phenylbutane, ksc523o6b |
PubChem CID | 9903372 |
IUPAC Name | tert-butyl N-[(1S)-1-[(2S)-oxiran-2-yl]-2-phenylethyl]carbamate |
SMILES | CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C2CO2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10021044
|
Thermo Scientific Acros
395020010 |
1 g | Glass bottle |
N/A
|
N/A | ||||
10366364
|
Thermo Scientific Acros
395020050 |
5 g | Glass bottle |
N/A
|
N/A | ||||
11985541
|
Thermo Scientific Acros
395020250 |
25 g | Glass bottle |
N/A
|
N/A | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
98737-29-2 | |
263.34 | |
NVPOUMXZERMIJK-QWHCGFSZSA-N | |
9903372 | |
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C2CO2 |
C15H21NO3 | |
MFCD02258997 | |
2s,3s-1,2-epoxy-3-boc-amino-4-phenylbutane, 2s,3s-n-t-boc-3-amino-1,2-epoxy-4-phenylbutane, tert-butyl s-1-s-oxiran-2-yl-2-phenylethyl carbamate, 2s,3s-1,2-epoxy-3-tert-butoxycarbonylamino-4-phenylbutane, tert-butyl n-1s-1-2s-oxiran-2-yl-2-phenylethyl carbamate, tert-butyl s-r*,r*---1-oxiranyl-2-phenylethyl carbamate, tert-butyl n-1s-1-2s-oxiran-2-yl-2-phenyl-ethyl carbamate, 2s,3s---3-t-boc-amino-1,2-epoxy-4-phenylbutane, ksc523o6b | |
tert-butyl N-[(1S)-1-[(2S)-oxiran-2-yl]-2-phenylethyl]carbamate |
Specifications
98737-29-2 | |
100.0 | |
97.5% min. (HPLC) | |
MFCD02258997 | |
Solubility in water: <0.1% (20°C) | |
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C2CO2 | |
263.34 | |
263.34 | |
(2S, 3S)-3-(N-BOC-amino)-1-oxirane-4-phenylbutane |
97.5 | |
125.0°C to 127.0°C | |
C15H21NO3 | |
2s,3s-1,2-epoxy-3-boc-amino-4-phenylbutane, 2s,3s-n-t-boc-3-amino-1,2-epoxy-4-phenylbutane, tert-butyl s-1-s-oxiran-2-yl-2-phenylethyl carbamate, 2s,3s-1,2-epoxy-3-tert-butoxycarbonylamino-4-phenylbutane, tert-butyl n-1s-1-2s-oxiran-2-yl-2-phenylethyl carbamate, tert-butyl s-r*,r*---1-oxiranyl-2-phenylethyl carbamate, tert-butyl n-1s-1-2s-oxiran-2-yl-2-phenyl-ethyl carbamate, 2s,3s---3-t-boc-amino-1,2-epoxy-4-phenylbutane, ksc523o6b | |
NVPOUMXZERMIJK-QWHCGFSZSA-N | |
tert-butyl N-[(1S)-1-[(2S)-oxiran-2-yl]-2-phenylethyl]carbamate | |
9903372 | |
98% |
Safety and Handling
GHS H Statement:
Very toxic to aquatic life with long lasting effects.
GHS P Statement:
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
- (2S,3S)-3-(N-BOC-amino)-1-oxirane-4-phenylbutane
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
Hazard Statement
- H410-Very toxic to aquatic life with long lasting effects.
- H400-Very toxic to aquatic life.
Precautionary Statement
- P273-Avoid release to the environment.
RUO – Research Use Only