missing translation for 'onlineSavingsMsg'
Learn More

(2S,3S)-3-Methylpyrrolidine-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog Number 10309450
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
10512-89-7
C6H11NO2
129.16
CNPSFBUUYIVHAP-WHFBIAKZSA-N
2s,3s-3-methylpyrrolidine-2-carboxylic acid, trans-3-methyl-l-proline, l-proline, 3-methyl-, 3s, trans-3-methylproline, 3beta-methyl-l-proline, 3s-3-methyl-l-proline, h-l-trans-pro 3-me-oh, 2s,3s-3-methylpyrrolidine-2-carboxylicacid, l-trans-3-methylproline, 2s,3s-3-methyl-pyrroline-2-carboxylic acid
2724879
(2S,3S)-3-methylpyrrolidine-2-carboxylic acid
CC1CCNC1C(=O)O
This item is not returnable. View return policy
10512-89-7
C6H11NO2
129.16
CNPSFBUUYIVHAP-WHFBIAKZSA-N
2s,3s-3-methylpyrrolidine-2-carboxylic acid, trans-3-methyl-l-proline, l-proline, 3-methyl-, 3s, trans-3-methylproline, 3beta-methyl-l-proline, 3s-3-methyl-l-proline, h-l-trans-pro 3-me-oh, 2s,3s-3-methylpyrrolidine-2-carboxylicacid, l-trans-3-methylproline, 2s,3s-3-methyl-pyrroline-2-carboxylic acid
2724879
(2S,3S)-3-methylpyrrolidine-2-carboxylic acid
CC1CCNC1C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 250 mg
Specific Rotation Condition − 28.00 (25.00°C c=0.7,water)
Specific Rotation − 28.00
Formula Weight 129.16
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material (2S, 3S)-3-Methylpyrrolidine-2-carboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • (2S,3S)-3-Methylpyrrolidine-2-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'