missing translation for 'onlineSavingsMsg'
Learn More

(2S,3S)-3-Boc-amino-1,2-epoxy-4-phenylbutane, TRC

Product Code. 30334633
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30334633 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30334633 Supplier Toronto Research Chemicals Supplier No. TRCB6185301G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material (2S,3S)-3-Boc-amino-1,2-epoxy-4-phenylbutane
CAS 98737-29-2
Molecular Formula C15 H21 N O3
Quantity 1 g
Synonym Carbamic acid, N-[(1S)-1-[(2S)-2-oxiranyl]-2-phenylethyl]-, 1,1-dimethylethyl ester, Carbamic acid, (1-oxiranyl-2-phenylethyl)-, 1,1-dimethylethyl ester, [S-(R*,R*)]-, Carbamic acid, [(1S)-1-(2S)-oxiranyl-2-phenylethyl]-, 1,1-dimethylethyl ester (9CI), (2S)-[(1S)-1-[(tert-Butoxycarbonyl)amino]-2-phenylethyl]oxirane, (2S,3S)-(-)-3-tert-Butoxycarbonylamino-1,2-epoxy-4-phenylbutane, (2S,3S)-3-[(tert-Butoxycarbonyl)amino]-1,2-epoxy-4-phenylbutane, [(1S)-1-((2S)-Oxiranyl)-2-phenylethyl]carbamic acid tert-butyl ester, tert-Butyl [(1S)-1-((2S)-oxiran-2-yl)-2-phenylethyl]carbamate, tert-Butyl [(1S)-1-((2S)-oxiranyl)-2-phenylethyl]carbamate, tert-Butyl [(1S)-1-[(2S)-oxiranyl]-2-phenylethyl]carbamate
InChI Formula InChI=1S/C15H21NO3/c1-15(2,3)19-14(17)16-12(13-10-18-13)9-11-7-5-4-6-8-11/h4-8,12-13H,9-10H2,1-3H3,(H,16,17)/t12-,13+/m0/s1
SMILES CC(C)(C)OC(=O)N[C@@H](Cc1ccccc1)[C@H]2CO2
IUPAC Name tert-butyl N-[(1S)-1-[(2S)-oxiran-2-yl]-2-phenylethyl]carbamate
Molecular Weight (g/mol) 263.33
Formula Weight 263.1521
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.