missing translation for 'onlineSavingsMsg'
Learn More

(2S,3S)-3-(Boc-amino)-1,2-epoxy-4-phenylbutane, 98%, Thermo Scientific™

Product Code. 15462406
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15462406 250 mg 250mg
15442406 1 g 1g
15452406 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15462406 Supplier Thermo Scientific Alfa Aesar Supplier No. H56001.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It function as a building block employed in the synthesis of HIV-1 protease inhibitors and is also employed in synthesizing (hydroxyethyl)urea peptidomimetrics and arylsulfonamides possessing anti-HIV activity. It is a Enantiomer S. Atazanavir intermediate.

Solubility
Insoluble in water. Soluble in Chloroform, Dichloromethane and Ethyl Acetate.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98737-29-2
Molecular Formula C15H21NO3
Molecular Weight (g/mol) 263.337
MDL Number MFCD02258997
InChI Key NVPOUMXZERMIJK-QWHCGFSZSA-N
Synonym 2s,3s-1,2-epoxy-3-boc-amino-4-phenylbutane, 2s,3s-n-t-boc-3-amino-1,2-epoxy-4-phenylbutane, tert-butyl s-1-s-oxiran-2-yl-2-phenylethyl carbamate, 2s,3s-1,2-epoxy-3-tert-butoxycarbonylamino-4-phenylbutane, tert-butyl n-1s-1-2s-oxiran-2-yl-2-phenylethyl carbamate, tert-butyl s-r*,r*---1-oxiranyl-2-phenylethyl carbamate, tert-butyl n-1s-1-2s-oxiran-2-yl-2-phenyl-ethyl carbamate, 2s,3s---3-t-boc-amino-1,2-epoxy-4-phenylbutane, ksc523o6b
PubChem CID 9903372
IUPAC Name tert-butyl N-[(1S)-1-[(2S)-oxiran-2-yl]-2-phenylethyl]carbamate
SMILES CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C2CO2

Specifications

Melting Point 125°C to 127°C
Quantity 250 mg
UN Number UN3077
Solubility Information Insoluble in water. Soluble in Chloroform,Dichloromethane and Ethyl Acetate.
Optical Rotation −71° (c=0.6 in Methanol)
Formula Weight 263.34
Percent Purity 98%
Chemical Name or Material (2S,3S)-3-(Boc-amino)-1,2-epoxy-4-phenylbutane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.