missing translation for 'onlineSavingsMsg'
Learn More

(2S,3S)-2-(4-Methoxyphenyl)-4-oxo-2,3,4,5-tetrahydro-1,5-benzothiazepin-3-yl Acetate, LoGiCal

Product Code. 30462467
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30462467 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30462467 Supplier LoGiCal Supplier No. LGCFOR0068.02

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2S,3S)-2-(4-Methoxyphenyl)-4-oxo-2,3,4,5-tetrahydro-1,5-benzothiazepin-3-yl Acetate
CAS 87447-47-0
Analyte or Component Names (2S,3S)-2-(4-Methoxyphenyl)-4-oxo-2,3,4,5-tetrahydro-1,5-benzothiazepin-3-yl Acetate
Shipping Condition Room Temperature
Molecular Formula C18 H17 N O4 S
Certifications/Compliance ISO 9001
Synonym Diltiazem Hydrochloride Imp. B (EP), Diltiazem Imp. B (EP), (2S,3S)-2-(4-Methoxyphenyl)-4-oxo-2,3,4,5-tetrahydro-1,5-benzothiazepin-3-yl Acetate, Diltiazem Hydrochloride Impurity B, Diltiazem Impurity B
InChI Formula InChI=1S/C18H17NO4S/c1-11(20)23-16-17(12-7-9-13(22-2)10-8-12)24-15-6-4-3-5-14(15)19-18(16)21/h3-10,16-17H,1-2H3,(H,19,21)/t16-,17+/m1/s1
Solution Type Neat
SMILES COc1ccc(cc1)[C@@H]2Sc3ccccc3NC(=O)[C@@H]2OC(=O)C
IUPAC Name [(2S,3S)-2-(4-methoxyphenyl)-4-oxo-3,5-dihydro-2H-1,5-benzothiazepin-3-yl] acetate
Industry Type Clinical Testing
Molecular Weight (g/mol) 343.4
Quantity 10 mg
Formula Weight 343.0878 g/mol
Show More Show Less
Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.