missing translation for 'onlineSavingsMsg'
Learn More

(2S,3S)-(+)-2,3-Butanediol, 99%

Product Code. 10276430 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
2.5 g
Unit Size:
2.5g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10276430 500 mg 500mg
10782675 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10276430 Supplier Thermo Scientific Acros Supplier No. 247735000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19132-06-0 | C4H10O2 | 90.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chiral derivatization reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19132-06-0
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD00063648
InChI Key OWBTYPJTUOEWEK-IMJSIDKUSA-N
Synonym 2s,3s-butane-2,3-diol, 2s,3s-+-2,3-butanediol, s,s-2,3-butanediol, s,s-+-2,3-butanediol, s,s-butane-2,3-diol, unii-7e9uxg71s1, 2,3-butanediol, +, s,s-2,3-butylene glycol, 2,3-butanediol, s-r*,r*, 2s,3s-2,3-butanediol
PubChem CID 439888
ChEBI CHEBI:16812
IUPAC Name (2S,3S)-butane-2,3-diol
SMILES CC(C(C)O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Density 0.9800g/mL
Boiling Point 179.0°C to 182.0°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(OH)CH(OH)CH3
Refractive Index 1.4309 to 1.4329
Quantity 500 mg
Beilstein 01, 2183
Fieser 01,82
Merck Index 15, 1571
Specific Gravity 0.98
Specific Rotation + 13.00
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and diisopropyl ether
Formula Weight 90.12
Percent Purity 99%
Physical Form Oily Liquid
Chemical Name or Material (2S, 3S)-(+)-2, 3-Butanediol, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.