missing translation for 'onlineSavingsMsg'
Learn More

(2S,3R,6R)-6-Ethyl-3-methyl-2-(3-oxobutyl)-cyclohexanone, TRC

Product Code. 30381194
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381194 5 mg -
30374336 1 mg 1mg
30386288 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381194 Supplier Toronto Research Chemicals Supplier No. TRCE6795605MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 958447-27-3
Molecular Formula C13H22O2
Molecular Weight (g/mol) 210.31
Synonym Dihydroartemisin Impurity H
IUPAC Name (2S,3R,6R)-6-ethyl-3-methyl-2-(3-oxobutyl)cyclohexan-1-one
SMILES CC[C@@H]1CC[C@@H](C)[C@H](CCC(=O)C)C1=O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C13H22O2/c1-4-11-7-5-9(2)12(13(11)15)8-6-10(3)14/h9,11-12H,4-8H2,1-3H3/t9-,11-,12+/m1/s1
Formula Weight 210.162
Chemical Name or Material (2S,3R,6R)-6-Ethyl-3-methyl-2-(3-oxobutyl)-cyclohexanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.