missing translation for 'onlineSavingsMsg'
Learn More

(2S)-3,4-Dihydro-1,2(2H)-pyridinedicarboxylic Acid 1,2-Bis(1,1-dimethylethyl) Ester, TRC

Product Code. 30387400
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387400 10 mg 10mg
30383241 5 mg 5mg
30377408 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387400 Supplier Toronto Research Chemicals Supplier No. TRCD45068510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2S)-3,4-Dihydro-1,2(2H)-pyridinedicarboxylic Acid 1,2-Bis(1,1-dimethylethyl) Ester
Analyte or Component Names (2S)-3,4-Dihydro-1,2(2H)-pyridinedicarboxylic Acid 1,2-Bis(1,1-dimethylethyl) Ester
Shipping Condition Room Temperature
Molecular Formula C15 H25 N O4
Synonym (2S)-3,4-Dihydro-1,2(2H)-pyridinedicarboxylic Acid Bis(1,1-dimethylethyl) Ester, (S)-Di-t-butyl 3,4-Dihydropyridine-1,2(2H)-dicarboxylate
InChI Formula InChI=1S/C15H25NO4/c1-14(2,3)19-12(17)11-9-7-8-10-16(11)13(18)20-15(4,5)6/h8,10-11H,7,9H2,1-6H3/t11-/m0/s1
Solution Type Neat
SMILES CC(C)(C)OC(=O)[C@@H]1CCC=CN1C(=O)OC(C)(C)C
IUPAC Name ditert-butyl (2S)-3,4-dihydro-2H-pyridine-1,2-dicarboxylate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 283.363
Quantity 10 mg
Formula Weight 283.178
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.