missing translation for 'onlineSavingsMsg'
Learn More

(2S)-2-Hydroxyglutaric Acid Disodium Salt, TRC

Product Code. 30392461
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392461 10 mg 10mg
30408708 5 mg 5mg
30401088 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30392461 Supplier Toronto Research Chemicals Supplier No. TRCH94257010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63512-50-5
Molecular Formula C5 H6 O5 . 2 Na
Molecular Weight (g/mol) 192.08
Synonym Pentanedioic acid, 2-hydroxy-, sodium salt (1:2), (2S)-, Pentanedioic acid, 2-hydroxy-, disodium salt, (S)- (9CI), (2S)-2-Hydroxypentanedioic acid disodium salt, (S)-(+)-2-Hydroxyglutaric acid disodium salt, (S)-2-Hydroxyglutaric acid disodium salt, (S)-2-Hydroxypentanedioic acid disodium salt, (S)-α-Hydroxyglutaric acid disodium salt, L-α-Hydroxyglutaric acid disodium salt
IUPAC Name disodium;(2S)-2-hydroxypentanedioate
SMILES [Na+].[Na+].O[C@@H](CCC(=O)[O-])C(=O)[O-]

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C5H8O5.2Na/c6-3(5(9)10)1-2-4(7)8;;/h3,6H,1-2H2,(H,7,8)(H,9,10);;/q;2*+1/p-2/t3-;;/m0../s1
Formula Weight 192.0011
Chemical Name or Material (2S)-2-Hydroxyglutaric Acid Disodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.